Learn More
Sino Biological Rat IL17F Protein 50ug (Sino Biological)
Supplier:  Sino Biological 80462R08B50
Encompass_Preferred
A DNA sequence encoding the rat Il17f (NP_001015011.1) (Met1-Ala153) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-1074