Learn More
Sino Biological Anti-IL-17RA Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80190T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat IL-17R / IL-17RA (CD217) (Catalog#80190-R08H; NP_001101353.2; Met1-Pro320). IL-17R / IL-17RA (CD217) specific IgG was purified by Rat IL-17R / IL-17RA (CD217) affinity chromatography.
Catalog No. 50-240-8938