Learn More
BIOHIPPO Rat IL19 Microsample
SDP
Catalog No. 5000314625
Encompass
Rat IL19 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL19 in Serum plasma and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 7 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat IL19 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat IL19 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat IL19 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14625 Supplier BIOHIPPO Supplier No. BHE15212137
Rat IL19 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL19 in Serum plasma and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 7 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat IL19 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat IL19 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat IL19 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color