Learn More
Sino Biological Recombinant Rat IL-1 alpha/IL1A Protein
Supplier:  Sino Biological 80385RNAE50
Encompass_Preferred
A DNA sequence encoding the rat IL1A (NP_058715.1) (Ser115-Ser270) was expressed.
Catalog No. 50-255-2775