Learn More
Sino Biological Recombinant Anti-IL-1 beta Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 80023R017100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat IL-1 beta (Catalog#80023-RNAE; Q63264-1; Val117-Ser268).
Catalog No. 50-241-0243