Learn More
Sino Biological Anti-IL-1 beta Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80023RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat IL-1 beta / IL1B (rR IL-1 beta / IL1B; Catalog#80023-RNAE; Q63264-1; Val 117-Ser 268). IL-1 beta / IL1B specific IgG was purified by Rat IL-1 beta / IL1B affinity chromatography.
Catalog No. 50-241-0787