Learn More
Sino Biological Anti-IL-1 beta Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80023RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat IL-1 beta (Catalog#80023-RNAE; Q63264-1; Val117-Ser268). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-0238