Learn More
Sino Biological Recombinant Anti-IL1R1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 80028R007100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat IL-1R1 / CD121a (rR IL-1R1 / CD121a; Catalog#80028-R08H; NP_037255.3; Met1-Lys352).
Catalog No. 50-240-9063