Learn More
Sino Biological Rat IL1R1 Matched ELISA Antibody Pair Set 15 Plates
Supplier: Sino Biological SEK8002815
The Rat IL-1R1 / CD121a ELISA Pair Set is for the quantitative determination of Rat IL-1R1 / CD121a.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for IL-1R1 / CD121a coated on a 96-well plate. Standards and samples are added to the wells, and any IL-1R1 / CD121a present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-IL-1R1 / CD121a monoclonal antibody is then added, producing an antibody-antigen-antibody “sandwich”. The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of IL-1R1 / CD121a present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.