Learn More
Sino Biological Rat IL1R2 Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000786374
Encompass_Preferred
The Rat IL-1R2 / CD121b ELISA Pair Set is for the quantitative determination of Rat IL-1R2 / CD121b.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Rat IL-1R2 / CD121b coated on a 96-well plate. Standards and samples are added to the wells, and any Rat IL-1R2 / CD121b present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Rat IL-1R2 / CD121b monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Rat IL-1R2 / CD121b present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86374 Supplier Sino Biological Supplier No. SEK8019615
The Rat IL-1R2 / CD121b ELISA Pair Set is for the quantitative determination of Rat IL-1R2 / CD121b.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Rat IL-1R2 / CD121b coated on a 96-well plate. Standards and samples are added to the wells, and any Rat IL-1R2 / CD121b present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Rat IL-1R2 / CD121b monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Rat IL-1R2 / CD121b present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.