Learn More
Sino Biological Rat IL2 MAb 25TS (Sino Biological)
Supplier:  Sino Biological 80075MM06A25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat IL2 (rR IL2; Catalog#80075-RNAE; P17108; Ala21-Gln155) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-237-1645