Learn More
Sino Biological Anti-IL-21 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80200RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat IL-21 (Catalog#80200-R08B; NP_001102413.1; Met1-Ser146). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2200