Learn More
Sino Biological Recombinant Rat IL22RA1 Protein (ECD, hFc Tag)
Supplier:  Sino Biological 80453R02H50
Encompass_Preferred
A DNA sequence encoding the rat Il22ra1 (NP_001178798.1) (Met1-Ala228) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-0728