Learn More
Sino Biological Recombinant Rat CD25/IL-2R alpha Protein (ECD, His & AVI Tag), Avi-tag Biotinylated
Catalog No. 502992747
Encompass_Preferred
A DNA sequence encoding the Rat IL2RA (NP_037295.1) (Met1-Gln235) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-299-2747 Supplier Sino Biological Supplier No. 80074R49BB100
A DNA sequence encoding the Rat IL2RA (NP_037295.1) (Met1-Gln235) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.