Learn More
Sino Biological Anti-CD25/IL-2R alpha Antibody, Mouse Monoclonal
Supplier:  Sino Biological 80074MM08100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat CD25/IL-2R alpha (rR CD25/IL-2R alpha; Catalog#80074-R08B; NP_037295.1; Met1-Gln235). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-2838