Learn More
Sino Biological Anti-IL-3 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 80235MM50100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat IL3 (rR IL3; Catalog#80235-R08B; NP_113701.1; Met1-Cys169). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-0730