Learn More
Sino Biological Recombinant Anti-IL-3 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 80235R210100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat IL3 (rR IL3; Catalog#80235-R08B; NP_113701.1; Met1-Cys169).
Catalog No. 50-240-9594