Learn More
Sino Biological Anti-IL-3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80235T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat IL3 / IL-3 / Interleukin-3 (rR IL3 / IL-3 / Interleukin-3; Catalog#80235-R08B; NP_113701.1; Met1-Cys169). IL3 / IL-3 / Interleukin-3 specific IgG was purified by Rat IL3 / IL-3 / Interleukin-3 affinity chromatography.
Catalog No. 50-240-9139