Learn More
Sino Biological Recombinant Rat IL-3 Protein (His Tag)
Supplier:  Sino Biological 80235R08B1
Encompass_Preferred
A DNA sequence encoding the rat IL3 (NP_113701.1) (Met 1-Cys 169) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-2806