Learn More
Sino Biological Recombinant Anti-IL-4R Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 80198R002A100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat IL4R (rR IL4R; Catalog#80198-R08H; Q63257-1; Met1-Arg232) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-241-0057