Learn More
Sino Biological Recombinant Anti-IL-4R Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 80198R002100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat IL-4R/CD124 (rR IL-4R/CD124; Catalog#80198-R08H; Q63257-1; Met1-Arg232).
Catalog No. 50-241-0434