Learn More
Sino Biological Rat IL6R Protein 100ug (Sino Biological)
Supplier:  Sino Biological 81072R02H100
Encompass_Preferred
A DNA sequence encoding the rat Il6r (NP_058716.2) (Met1-Pro364) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-2687