Learn More
Sino Biological Recombinant Anti-gp130/IL6ST Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 80030R108P100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat gp130 / CD130 / IL6ST (rR gp130 / CD130 / IL6ST; Catalog#80030-R08H; NP_001008725.2; Met4-Glu618) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-255-2861