Learn More
Sino Biological Anti-gp130/IL6ST Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80030RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat IL6ST / gp130 / CD130 (Catalog#80030-R08H; NP_001008725.2; Met4-Glu618). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-2859