Learn More
Sino Biological Recombinant Anti-IL-7 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 80329R075100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat IL7 / IL-7 / Interleukin-7 (rR IL7 / IL-7 / Interleukin-7; Catalog#80329-RNAE; Q91Y32; Asp26-Ile154).
Catalog No. 50-240-7662