Learn More
BIOHIPPO Rat IL7R Microsample
SDP
Catalog No. 5000313756
Encompass
Rat IL7R Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL7R in Serum plasma and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 122 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat IL7R Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat IL7R Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat IL7R biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13756 Supplier BIOHIPPO Supplier No. BHE15211235
Rat IL7R Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL7R in Serum plasma and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 122 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat IL7R Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat IL7R Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat IL7R biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color