Learn More
Sino Biological Recombinant Anti-IL-9 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 80459R030100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat IL-9 (rR IL-9; Catalog#80459-R08B; NP_001099217.1; Met1-Ala144).
Catalog No. 50-241-2574