Learn More
Sino Biological Anti-IL-9 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 80459MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat IL-9 (rR IL-9; Catalog#80459-R08B; NP_001099217.1; Met1-Ala144). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1550