Learn More
Sino Biological Rat IL9 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 80459T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat IL-9 (Catalog#80459-R08B; NP_001099217.1; Met1-Ala144). IL-9 specific IgG was purified by Rat IL-9 affinity chromatography.
Catalog No. 50-237-2958