Learn More
Sino Biological Rat ILT11/LILRA5/LIR-9 Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000785837
Encompass_Preferred
The Rat LILRA5 ELISA Pair Set is for the quantitative determination of Rat LILRA5.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Rat LILRA5 coated on a 96-well plate. Standards and samples are added to the wells, and any Rat LILRA5 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Rat LILRA5 polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Rat LILRA5 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85837 Supplier Sino Biological Supplier No. SEK8033415
The Rat LILRA5 ELISA Pair Set is for the quantitative determination of Rat LILRA5.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Rat LILRA5 coated on a 96-well plate. Standards and samples are added to the wells, and any Rat LILRA5 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Rat LILRA5 polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Rat LILRA5 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.