Learn More
BIOHIPPO Rat IMA Microsample
SDP
Catalog No. 5000314604
Encompass
Rat IMA Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IMA in serum plasma and other biological fluids It provides a detection range of 6 25-400 ng/mL with a sensitivity of 3 68 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat IMA Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat IMA Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat IMA biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14604 Supplier BIOHIPPO Supplier No. BHE15210716
Rat IMA Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IMA in serum plasma and other biological fluids It provides a detection range of 6 25-400 ng/mL with a sensitivity of 3 68 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat IMA Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat IMA Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat IMA biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color