Learn More
Biomatik Corporation Rat Immunoglobulin D (IgD) ELISA Kit
Supplier:  Biomatik Corporation EKU08733
Encompass_Preferred
Rat Immunoglobulin D (IgD) ELISA Kit for detecting IgD in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 123.46-10000pg/ml. Sensitivity: 53.7pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01883. Target Synonyms: IGHD; Constant Region Of Heavy Chain Of Immunoglobulin Delta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8633