Learn More
Biomatik Corporation Rat Immunoglobulin G (IgG) CLIA Kit
Supplier:  Biomatik Corporation EKU08635
Encompass_Preferred
Rat Immunoglobulin G (IgG) CLIA Kit for detecting IgG in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 781-200,000ng/ml. Sensitivity: 265ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8649