Learn More
Biomatik Corporation Rat Immunoglobulin G (IgG) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10933
Encompass_Preferred
Rat Immunoglobulin G (IgG) High-sensitive ELISA Kit for detecting IgG in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.8-500ng/ml. Sensitivity: 3.5ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8266