Learn More
Biomatik Corporation Rat Immunoglobulin G2a (IgG2a) ELISA Kit
Supplier:  Biomatik Corporation EKU04906
Encompass_Preferred
Rat Immunoglobulin G2a (IgG2a) ELISA Kit for detecting IgG2a in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 246.91-20000ng/ml. Sensitivity: 96.3ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P20760. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8927