Learn More
American Research Products Inc Rat Indoleamine 2 3 dioXygena
Catalog No. 5000328399
Encompass
Rat Indoleamine 2 3-dioXygenase IDO ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28399 Supplier American Research Products Inc Supplier No. CSBE13583R
Rat Indoleamine 2 3-dioXygenase IDO ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 81 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich