Learn More
American Research Products Inc Rat inducible nitric oXide syn
Catalog No. 5000329706
Encompass
Rat inducible nitric oXide synthase iNOS ELISA KIT from CUSABIO Detection Range 0 78 IU/mL-50 IU/mL Sensitivity 0 195 IU/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29706 Supplier American Research Products Inc Supplier No. CSBE08325R
Rat inducible nitric oXide synthase iNOS ELISA KIT from CUSABIO Detection Range 0 78 IU/mL-50 IU/mL Sensitivity 0 195 IU/mL Sample serum plasma tissue homogenates Method Sandwich