Learn More
Raybiotech Inc G-Series Rat Inflammation Array 1 Kit.
Catalog No. 501950504
Encompass_Preferred
Semi-quantitative measurement of 10 rat cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-2, IL-4, IL-6, IL-10, IL-13, MCP-1 (CCL2), TNF alpha
Catalog No. 50-195-0504 Supplier Raybiotech Inc Supplier No. GSRINF11
Semi-quantitative measurement of 10 rat cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-2, IL-4, IL-6, IL-10, IL-13, MCP-1 (CCL2), TNF alpha