Learn More
Raybiotech Inc Quantibody Rat Inflammation Array 1 Kit.
Catalog No. 501950911
Encompass_Preferred
Detects 10 Rat Inflammatory Factors. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-2, IL-4, IL-6, IL-10, IL-13, MCP-1 (CCL2), TNF alpha
Catalog No. 50-195-0911 Supplier Raybiotech Inc Supplier No. QARINF11
Detects 10 Rat Inflammatory Factors. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: IFN-gamma, IL-1 alpha (IL-1 F1), IL-1 beta (IL-1 F2), IL-2, IL-4, IL-6, IL-10, IL-13, MCP-1 (CCL2), TNF alpha