Learn More
Biomatik Corporation Rat Inhibin Alpha (INHa) ELISA Kit
Supplier:  Biomatik Corporation EKU10138
Encompass_Preferred
Rat Inhibin Alpha (INHa) ELISA Kit for detecting INHa in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.15pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P17490. Gene ID: 24504. Target Synonyms: INH-A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6069