Learn More
Biomatik Corporation Rat Inhibin Beta C (INHbC) ELISA Kit
Supplier:  Biomatik Corporation EKU04938
Encompass_Preferred
Rat Inhibin Beta C (INHbC) ELISA Kit for detecting INHbC in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.12-5000pg/ml. Sensitivity: 25pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9WUK5. Gene ID: 64549. Target Synonyms: IHBC; INH-Bc. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7392