Learn More
American Research Products Inc Rat inositol 1 4 5 trispho
Catalog No. 5000329270
Encompass
Rat inositol 1 4 5 -trisphosphate (IP3) ELISA kit from CUSABIO Detection Range 5 pg/mL-1000 pg/mL Sensitivity 2 5 pg/mL Sample serum plasma tissue homogenates cell culture supernates cell lysates Method Competitive
Catalog No. 50-003-29270 Supplier American Research Products Inc Supplier No. CSBE13004R
Rat inositol 1 4 5 -trisphosphate (IP3) ELISA kit from CUSABIO Detection Range 5 pg/mL-1000 pg/mL Sensitivity 2 5 pg/mL Sample serum plasma tissue homogenates cell culture supernates cell lysates Method Competitive