Learn More
BIOHIPPO Rat INS Microsample
SDP
Catalog No. 5000315571
Encompass
Rat INS Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of INS in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 052 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat INS Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat INS Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat INS biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15571 Supplier BIOHIPPO Supplier No. BHE15212615
Rat INS Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of INS in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 052 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat INS Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat INS Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat INS biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color