Learn More
Proteintech Group Inc Rat Insulin ELISA Kit 96well
SDP
Supplier:  Proteintech Group Inc KE2000896T
Encompass_Preferred
Insulin is a peptide hormone, produced by beta cells of the pancreas, and is central to regulating carbohydrate and fat metabolism in the body. It participates in glucose utilization, protein synthesis and in the formation and storage of neutral lipids. Insulin is synthesized as a precursor molecule, proinsulin, which is processed prior to secretion. A- and B-peptides are joined together by a disulfide bond to form insulin, while the central portion of the precursor molecule is cleaved and released as the C-peptide. Defects in insulin results in type 1 diabetes mellitus.
Catalog No. 50-197-4180