Learn More
Crystal Chem Inc Rat Insulin ELISA KIT, 96-wells
SDP
Supplier:  Crystal Chem Inc 90010
Encompass
The Rat Insulin ELISA is used to quantify levels of rat insulin in serum, plasma, and fluid using only a 5 µL sample volume. Highlights: Kits use a sample size of only 5 µL. Wide dynamic range of 0.156 to 10.0 ng/mL. High Sensitivity of 0.156 ng/mL. We also offer an Ultra Sensitive Rat Insulin kit. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7913
Edge