Learn More
Biomatik Corporation Rat Insulin Like Growth Factor Binding Protein 6 (IGFBP6) ELISA Kit
Supplier:  Biomatik Corporation EKU05007
Encompass_Preferred
Rat Insulin Like Growth Factor Binding Protein 6 (IGFBP6) ELISA Kit for detecting IGFBP6 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.066ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35572. Gene ID: 25641. Target Synonyms: IBP6; IGF-binding protein 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7553