Learn More
American Research Products Inc Rat Insulin like growth factor
Supplier:  American Research Products Inc CSBE04582R
Encompass
Rat Insulin-like growth factor 1 IGF-1 ELISA Kit from CUSABIO Detection Range 0 156 ng/ml-10 ng/ml Sensitivity 0 151 ng/ml Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26441