Learn More
American Research Products Inc Rat Insulin like growth factor
Catalog No. 5000329899
Encompass
Rat Insulin-like growth factor 2 IGF-2 ELISA Kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29899 Supplier American Research Products Inc Supplier No. CSBE04585R
Rat Insulin-like growth factor 2 IGF-2 ELISA Kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates Method Sandwich