Learn More
American Research Products Inc Rat Insulin like growth factor
Catalog No. 5000329597
Encompass
Rat Insulin-like growth factor binding protein 1 IGFBP-1 ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29597 Supplier American Research Products Inc Supplier No. CSBE07349R
Rat Insulin-like growth factor binding protein 1 IGFBP-1 ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich