Learn More
American Research Products Inc Rat Insulin like growth factor
Supplier:  American Research Products Inc CSBE07351R
Encompass
Rat Insulin-like growth factor binding protein 2 IGFBP-2 ELISA kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma tissuehomogenates Method Sandwich
Catalog No. 50-003-26460